Learn More
Metro™ Wire Shelving Track Sets
Product Code. 15987311
Change view
Click to view available options
Length (English):
27 in.
72 in.
84 in.
96 in.
108 in.
120 in.
132 in.
144 in.
156 in.
168 in.
180 in.
192 in.
204 in.
216 in.
228 in.
240 in.
252 in.
Unit Size:
Each
Product Code. Length (English) unitSize
15987311 27 in. Each
15937321 96 in. Each
15947321 108 in. Each
15997311 240 in. Each
15917321 72 in. Each
15987301 120 in. Each
15937311 180 in. Each
15997301 132 in. Each
15917311 156 in. Each
15947311 192 in. Each
15977311 228 in. Each
15967311 216 in. Each
15927321 84 in. Each
15907321 252 in. Each
15907311 144 in. Each
15927311 168 in. Each
15957311 204 in. Each
17 options
Product Code. 15987311
Brand: Metro™ TTS2.5NA
Only null left
Add to basket
Add to basket
For use with Metro Super Erecta™, Super Erecta Pro™ and Super Adjustable™ Top Track™ shelving.
  • Track sets include necessary sections of track for assembling track runs up to 21 ft. (640 cm)
  • Coated with Microban™ antimicrobial protection
Order Info
19-138-228 (TTS2.5NA) includes a joining kit to connect the track to other track sets.
Specifications
Product Type Track Set
For Use With (Equipment) Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ top track shelving
Includes Necessary sections of track for assembling track runs of 1830 to 6405mm
Length (English) 27 in.
Length (Metric) 76.2 cm