Learn More
Metro™ Super Erecta™ Wire Shelf, Stainless Steel Finish
Product Code. 15956701
Change view
Click to view available options
Dimensions (L x W):
35.5 x 121.9 in. (14 x 48 cm)
35.5 x 182.9 in. (14 x 72 cm)
36 x 60 in. (91 x 152.4 cm)
45.7 x 121.9 in. (18 x 48 cm)
45.7 x 183 in. (18 x 72 cm)
45.7 x 61 in. (18 x 24 cm)
45.7 x 91.4 in. (18 x 36 cm)
53 x 106.6 in. (21 x 42 cm)
53 x 21.9 in. (21 x 48 cm)
53 x 37 in. (21 x 54 cm)
53 x 63 in. (28 x 28 cm)
53 x 76 in. (30 x 30 cm)
61 x 106.6 in. (24 x 42 cm)
61 x 121.9 in. (24 x 48 cm)
61 x 137 in. (24 x 54 cm)
61 x 152.4 in. (24 x 60 cm)
76 x 121.9 in. (30 x 48 cm)
76 x 182.9 in. (30 x 72 cm)
76 x 91.4 in. (30 x 36 cm)
91 x 182.9 in. (36 x 72 cm)
91 x 91 in. (36 x 36 cm)
Depth (Metric):
355 mm
457 mm
530 mm
610 mm
760 mm
910 mm
Unit Size:
Each
Product Code. Dimensions (L x W) Depth (Metric) unitSize
15956701 36 x 60 in. (91 x 152.4 cm) 910 mm Each
15946611 45.7 x 183 in. (18 x 72 cm) 457 mm Each
15966681 76 x 121.9 in. (30 x 48 cm) 760 mm Each
15946681 76 x 91.4 in. (30 x 36 cm) 760 mm Each
15916671 61 x 121.9 in. (24 x 48 cm) 610 mm Each
15926591 45.7 x 61 in. (18 x 24 cm) 457 mm Each
15976671 61 x 152.4 in. (24 x 60 cm) 610 mm Each
15966601 45.7 x 121.9 in. (18 x 48 cm) 457 mm Each
15906601 45.7 x 91.4 in. (18 x 36 cm) 457 mm Each
15906581 35.5 x 121.9 in. (14 x 48 cm) 355 mm Each
15986581 35.5 x 182.9 in. (14 x 72 cm) 355 mm Each
15996611 53 x 63 in. (28 x 28 cm) 530 mm Each
15986631 53 x 21.9 in. (21 x 48 cm) 530 mm Each
15946621 53 x 76 in. (30 x 30 cm) 530 mm Each
15936631 53 x 106.6 in. (21 x 42 cm) 530 mm Each
15916641 53 x 37 in. (21 x 54 cm) 530 mm Each
15986661 61 x 106.6 in. (24 x 42 cm) 610 mm Each
15946671 61 x 137 in. (24 x 54 cm) 610 mm Each
15926691 76 x 182.9 in. (30 x 72 cm) 760 mm Each
15916701 91 x 91 in. (36 x 36 cm) 910 mm Each
15986701 91 x 182.9 in. (36 x 72 cm) 910 mm Each
21 options
Product Code. 15956701
Brand: Metro™ 3660NS
Wire shelves in stainless steel finish allow adjustable configurations when used with Metro Super Erecta shelving systems.
  • Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
  • Type 304 polished stainless steel
  • Used for the most aggressive applications and environments
  • Stands up to high temperature automated cart wash and autoclave applications
Specifications
Product Type Wire Shelf
Material Stainless Steel
Depth (English) 36 in.
Depth (Metric) 910 mm
Dimensions (L x W) 36 x 60 in. (91 x 152.4 cm)
For Use With (Equipment) Metro Super Erecta Shelving System
Includes Plastic sleeves and wedges