Learn More
Metro™ Super Erecta™ Wire Shelf, Metroseal 3 with Microban™ Finish
Product Code. 15986681
Change view
Click to view available options
Dimensions (L x W):
14 x 30 in.
14 x 36 in.
14 x 48 in.
14 x 60 in.
14 x 72 in.
18 x 24 in.
18 x 30 in.
18 x 36 in.
18 x 54 in.
18 x 60 in.
21 x 24 in.
21 x 30 in.
21 x 36 in.
21 x 42 in.
21 x 48 in.
21 x 54 in.
21 x 72 in.
24 x 24 in.
24 x 30 in.
24 x 36 in.
24 x 42 in.
24 x 48 in.
24 x 54 in.
30 x 48 in.
30 x 60 in.
30 x 72 in.
36 x 48 in.
36 x 60 in.
36 x 72 in.
Unit Size:
Each
29 product options available for selection
Product selection table with 29 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
15986681 30 x 60 in. Each
15906571 14 x 30 in. Each
15946571 14 x 36 in. Each
15996571 14 x 48 in. Each
15946581 14 x 60 in. Each
15976581 14 x 72 in. Each
15916591 18 x 24 in. Each
15956591 18 x 30 in. Each
15996591 18 x 36 in. Each
15986601 18 x 54 in. Each
15926611 18 x 60 in. Each
15986611 21 x 24 in. Each
15936621 21 x 30 in. Each
15926631 21 x 42 in. Each
15986621 21 x 36 in. Each
15976631 21 x 48 in. Each
15906641 21 x 54 in. Each
15926651 24 x 24 in. Each
15976651 24 x 30 in. Each
15986641 21 x 72 in. Each
15926661 24 x 36 in. Each
15976661 24 x 42 in. Each
15936671 24 x 54 in. Each
15906671 24 x 48 in. Each
15956681 30 x 48 in. Each
15916691 30 x 72 in. Each
15936701 36 x 48 in. Each
15946701 36 x 60 in. Each
15976701 36 x 72 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 29 options available.
29 options
Product Code. 15986681 Supplier Metro™ Supplier No. 3060NK3
Wire shelves are easily adjustable and the Metroseal 3 green epoxy with Microban™ finish promotes easy cleaning. For use with Metro Super Erecta shelving systems.
  • Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
  • Corrosion resistant finish for wet or high humidity environments
  • Antimicrobial is built into finish to keep product clean
Specifications
Product Type Wire Shelf
Dimensions (L x W) 30 x 60 in.
Finish Metroseal 3 with Microban