Learn More
Metro™ Super Erecta™ Wire Shelf, Metroseal 3 with Microban™ Finish
Product Code. 15916591
Change view
Click to view available options
Dimensions (L x W):
14 x 30 in.
14 x 36 in.
14 x 48 in.
14 x 60 in.
14 x 72 in.
18 x 24 in.
18 x 30 in.
18 x 36 in.
18 x 54 in.
18 x 60 in.
21 x 24 in.
21 x 30 in.
21 x 36 in.
21 x 42 in.
21 x 48 in.
21 x 54 in.
21 x 72 in.
24 x 24 in.
24 x 30 in.
24 x 36 in.
24 x 42 in.
24 x 48 in.
24 x 54 in.
30 x 48 in.
30 x 60 in.
30 x 72 in.
36 x 48 in.
36 x 60 in.
36 x 72 in.
Unit Size:
Each
| Product Code. | Dimensions (L x W) | unitSize |
|---|---|---|
| 15916591 | 18 x 24 in. | Each |
| 15906571 | 14 x 30 in. | Each |
| 15946571 | 14 x 36 in. | Each |
| 15996571 | 14 x 48 in. | Each |
| 15946581 | 14 x 60 in. | Each |
| 15976581 | 14 x 72 in. | Each |
| 15956591 | 18 x 30 in. | Each |
| 15996591 | 18 x 36 in. | Each |
| 15986601 | 18 x 54 in. | Each |
| 15926611 | 18 x 60 in. | Each |
| 15986611 | 21 x 24 in. | Each |
| 15936621 | 21 x 30 in. | Each |
| 15926631 | 21 x 42 in. | Each |
| 15986621 | 21 x 36 in. | Each |
| 15976631 | 21 x 48 in. | Each |
| 15906641 | 21 x 54 in. | Each |
| 15926651 | 24 x 24 in. | Each |
| 15976651 | 24 x 30 in. | Each |
| 15986641 | 21 x 72 in. | Each |
| 15926661 | 24 x 36 in. | Each |
| 15976661 | 24 x 42 in. | Each |
| 15936671 | 24 x 54 in. | Each |
| 15906671 | 24 x 48 in. | Each |
| 15986681 | 30 x 60 in. | Each |
| 15956681 | 30 x 48 in. | Each |
| 15916691 | 30 x 72 in. | Each |
| 15936701 | 36 x 48 in. | Each |
| 15946701 | 36 x 60 in. | Each |
| 15976701 | 36 x 72 in. | Each |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
- Corrosion resistant finish for wet or high humidity environments
- Antimicrobial is built into finish to keep product clean
Specifications
Specifications
| Product Type | Wire Shelf |
| Dimensions (L x W) | 18 x 24 in. |
| Finish | Metroseal 3 with Microban |