Learn More
Metro™ Super Erecta™ Top Track Stationary Intermediate Unit Kits
Description
Includes
- 2185mm (86 in.) posts
- Hardware for connecting stationary intermediate unit to tracks on both sides
Top-Track storage systems increase storage capacity and ease of access with overhead guide tracks. This design allows for open aisles eliminating hard to reach spaces, an interchangeable/modular storage system and easier cleaning.
- Stationary intermediate unit helps organize Super Erecta, Super Erecta Pro™ and Super Adjustable™ Top-Track storage systems
- 2185mm (86 in.) high posts
MetroSeal 3 Finish
Is a multi-layer corrosion resistant finish consisting of a proprietary cross linked thermoset epoxy enhanced with Microban antimicrobial protection over a zinc chromate coating. Metroseal 3 with Microban provides protection from bacteria, mold, mildew, and fungi build-up that may cause odor, stains, and product degradation, while displaying excellent abrasion, gouge, and chemical resistance properties.
Order Info
Shelves are ordered separately
Specifications
Specifications
| Product Type | Stationary Intermediate Unit Kit |
| Finish | Metroseal 3 with Microban™ |
| For Use With (Equipment) | Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ Top-Track™ storage system |
| Height (English) | 86 in. |
| Height (Metric) | 2185 mm |
| Includes | Posts, casters/channel caster, donut bumpers and roller bearing assemblies, hardware, split sleeves |
| Weight (English) | 8 lb. |
| Weight (Metric) | 3.6 kg |
| Width (English) Shelf | 21 in. |
| Width (Metric) Shelf | 53 cm |
| Show More |