Learn More
Metro™ Super Erecta™ Top Track Stationary Intermediate Unit Kits
Product Code. 15937281
Change view
Click to view available options
Material:
Stainless Steel
Steel
Width (English) Shelf:
18 in.
21 in.
24 in.
Finish:
Chrome
Metroseal 3 with Microban™
None
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Width (English) Shelf Finish unitSize
15937281 Stainless Steel 18 in. None Each
15967281 Stainless Steel 21 in. None Each
15997281 Stainless Steel 24 in. None Each
15917281 Steel 18 in. Chrome Each
15947281 Steel 21 in. Chrome Each
15977281 Steel 24 in. Chrome Each
15927281 Steel 18 in. Metroseal 3 with Microban™ Each
15957281 Steel 21 in. Metroseal 3 with Microban™ Each
15987281 Steel 24 in. Metroseal 3 with Microban™ Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15937281 Supplier Metro™ Supplier No. TTA18S
  • ONLINE EXCLUSIVE PRICE
    6520.00 NOK
    5738.65 NOK / Pack of 1
    Save 781.35 NOK
    11% Off
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
Includes
  • 2185mm (86 in.) posts
  • Hardware for connecting stationary intermediate unit to tracks on both sides
High-density, easy access storage
Top-Track storage systems increase storage capacity and ease of access with overhead guide tracks. This design allows for open aisles eliminating hard to reach spaces, an interchangeable/modular storage system and easier cleaning.
  • Stationary intermediate unit helps organize Super Erecta, Super Erecta Pro™ and Super Adjustable™ Top-Track storage systems
  • 2185mm (86 in.) high posts
Order Info
Shelves are ordered separately
Specifications
Product Type Stationary Intermediate Unit Kit
Finish None
For Use With (Equipment) Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ Top-Track™ storage system
Height (English) 86 in.
Height (Metric) 2185 mm
Includes Posts, casters/channel caster, donut bumpers and roller bearing assemblies, hardware, split sleeves
Weight (English) 7.5 lb.
Weight (Metric) 3.3 kg
Width (English) Shelf 18 in.
Width (Metric) Shelf 45.7 cm
Material Stainless Steel