Learn More
Metro™ Super Erecta™ Top Track Mobile Unit Kits
Description
Includes
Posts, casters/caster channels, donut bumpers and roller bearing assemblies
Top-Track storage systems increase storage capacity and ease of access with overhead guide tracks. This design allows for open aisles eliminating hard to reach spaces, an interchangeable/modular storage system and easier cleaning.
- Mobile units move along anchored track allowing easy access to all shelves
MetroSeal 3 Finish
Is a multi-layer corrosion resistant finish consisting of a proprietary cross linked thermoset epoxy enhanced with Microban antimicrobial protection over a zinc chromate coating. Metroseal 3 with Microban provides protection from bacteria, mold, mildew, and fungi build-up that may cause odor, stains, and product degradation, while displaying excellent abrasion, gouge, and chemical resistance properties.
Order Info
Shelves are ordered separately. One kit required per mobile unit.
Specifications
Specifications
| Product Type | Mobile Unit Kit |
| Finish | Metroseal 3 with Microban™ |
| For Use With (Equipment) | Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ Top-Track™ storage system |
| Includes | Posts, casters/channel caster, donut bumpers and roller bearing assemblies, hardware, split sleeves |
| Weight (English) | 32 lb. |
| Weight (Metric) | 14.4 kg |
| Width (English) Shelf | 18 in. |
| Width (English) | 20.25 in. |
| Width (Metric) Shelf | 45.7 cm |
| Width (Metric) | 51.4 cm |
| Show More |