Learn More
Metro™ Super Erecta™, Super Erecta Pro™ and Super Adjustable™ 2 Top-Track Stationary End Unit Kits
Product Code. 15967291
Change view
Click to view available options
Material:
Stainless Steel
Steel
Width (English) Shelf:
18 in.
21 in.
24 in.
Finish:
Chrome
Metroseal 3 with Microban™
Metroseal Green Epoxy
None
Unit Size:
Each
Product Code. Material Width (English) Shelf Finish unitSize
15967291 Steel 24 in. Chrome Each
15927291 Stainless Steel 18 in. None Each
15957291 Stainless Steel 21 in. None Each
15987291 Stainless Steel 24 in. None Each
15907291 Steel 18 in. Chrome Each
15937291 Steel 21 in. Chrome Each
15917291 Steel 18 in. Metroseal 3 with Microban™ Each
15947291 Steel 21 in. Metroseal 3 with Microban™ Each
15977291 Steel 24 in. Metroseal Green Epoxy Each
9 options
Product Code. 15967291
Brand: Metro™ TTE24C
Connects stationary end units to overhead track sets.
  • Kit includes 86 in. (218.5cm) posts and hardware necessary for connecting stationary end units to track
  • Super Erecta Shelves open-wire design minimizes dust accumulation, allows a free circulation of air and greater visibility of stored items.
Order Info
One kit is requird per single system. Shelves sold separately.
Specifications
Product Type Stationary End Unit Kit
Finish Chrome
For Use With (Equipment) Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ Top-Track™ storage system
Includes 86 in. Posts and connecting hardware
Weight (English) 12 lb.
Weight (Metric) 5.4 kg
Width (English) Shelf 24 in.
Width (English) 24 in.
Width (Metric) Shelf 61 cm
Width (Metric) 61 cm
Material Steel