Learn More
Metro™ Super Erecta™ Solid Shelf, Stainless Steel, Autoclavable and Cart-Washable
Description
- All-stainless construction
- Order aluminum split sleeves with stainless C-rings for corrosive, high-temperature autoclave or cart washing applications (13-450-953)
Specifications
Specifications
| Product Type | Solid Shelf |
| Color | Silver |
| Material | Stainless Steel |
| Autoclavable | Autoclavable |
| Dimensions (L x W) | 48 x 21 in. (121.9 x 53 cm) |
| For Use With (Equipment) | Metro Super Erecta Shelving System |
| Length (English) | 48 in. |
| Length (Metric) | 121.9 cm |
| Width (English) | 21 in. |
| Width (Metric) | 53 cm |