Learn More
Metro™ Super Erecta™ Solid Shelf, Stainless Steel, Autoclavable and Cart-Washable
Product Code. 15936581
Change view
Click to view available options
Width (English):
14 in.
18 in.
21 in.
24 in.
Length (English):
24 in.
30 in.
36 in.
42 in.
48 in.
60 in.
Unit Size:
Each
Product Code. Width (English) Length (English) unitSize
15936581 14 in. 60 in. Each
15966651 24 in. 30 in. Each
15976571 14 in. 42 in. Each
15946591 18 in. 30 in. Each
15926601 18 in. 42 in. Each
15916611 18 in. 60 in. Each
15976611 21 in. 24 in. Each
15916631 21 in. 42 in. Each
15966631 21 in. 48 in. Each
15966661 24 in. 42 in. Each
15986571 14 in. 48 in. Each
15996561 14 in. 30 in. Each
15966671 24 in. 60 in. Each
15916661 - - Each
15926621 21 in. 30 in. Each
15986561 14 in. 24 in. Each
15936571 14 in. 36 in. Each
15986591 18 in. 36 in. Each
15956601 18 in. 48 in. Each
15996661 24 in. 48 in. Each
15906591 18 in. 24 in. Each
15976621 21 in. 36 in. Each
15946641 - - Each
23 options
Product Code. 15936581
Brand: Metro™ 1460NFS
Keep autoclave equipment neat and in place. Metro Super Erecta™ Solid Shelf is perfect for autoclave and cart washing applications.
  • All-stainless construction
  • Order aluminum split sleeves with stainless C-rings for corrosive, high-temperature autoclave or cart washing applications (13-450-953)
Specifications
Product Type Solid Shelf
Color Silver
Material Stainless Steel
Autoclavable Autoclavable
Dimensions (L x W) 60 x 14 in. (152.4 x 35.5 cm)
For Use With (Equipment) Metro Super Erecta Shelving System
Length (English) 60 in.
Length (Metric) 152.4 cm
Width (English) 14 in.
Width (Metric) 35.5 cm