Learn More
Metro™ Super Erecta™ Solid Shelf, Stainless Steel, Autoclavable and Cart-Washable
Product Code. 15916661
Change view
Click to view available options
Unit Size:
Each
Width (English):
14 in.
18 in.
21 in.
24 in.
Length (English):
24 in.
30 in.
36 in.
42 in.
48 in.
60 in.
Product Code. unitSize Width (English) Length (English)
15916661 Each - -
15966651 Each 24 in. 30 in.
15976571 Each 14 in. 42 in.
15946591 Each 18 in. 30 in.
15936581 Each 14 in. 60 in.
15926601 Each 18 in. 42 in.
15916611 Each 18 in. 60 in.
15976611 Each 21 in. 24 in.
15916631 Each 21 in. 42 in.
15966631 Each 21 in. 48 in.
15966661 Each 24 in. 42 in.
15986571 Each 14 in. 48 in.
15996561 Each 14 in. 30 in.
15966671 Each 24 in. 60 in.
15926621 Each 21 in. 30 in.
15986561 Each 14 in. 24 in.
15936571 Each 14 in. 36 in.
15986591 Each 18 in. 36 in.
15956601 Each 18 in. 48 in.
15996661 Each 24 in. 48 in.
15906591 Each 18 in. 24 in.
15976621 Each 21 in. 36 in.
15946641 Each - -
23 options
Product Code. 15916661
Brand: Metro™ 2436NFS
Keep autoclave equipment neat and in place. Metro Super Erecta™ Solid Shelf is perfect for autoclave and cart washing applications.
  • All-stainless construction
  • Order aluminum split sleeves with stainless C-rings for corrosive, high-temperature autoclave or cart washing applications (13-450-953)
Specifications
Product Type Solid Shelf
Color Silver
Material Stainless Steel
Autoclavable Autoclavable
Dimensions (L x W) 36 x 24 in. (91.4 x 61 cm)
For Use With (Equipment) Metro Super Erecta Shelving System
Width (Metric) 61 cm