Learn More
Metro™ Super Erecta™ Solid Shelves, Stainless Steel, Autoclavable and Cart-Washable
Product Code. 15916651
Change view
Click to view available options
Length (English):
24 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Unit Size
15916651 24 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15916651 Supplier Metro™ Supplier No. 2424NFS
  • ONLINE EXCLUSIVE PRICE
    4120.00 NOK
    3828.34 NOK / Pack of 1
    Save 291.66 NOK
    7% Off
Estimated Shipment: 04-03-2027
For use with Metro™ Super Erecta™ shelving system
  • All-stainless construction
Order aluminum split sleeves with stainless C-rings for corrosive, high-temperature autoclave or cart washing applications (Fisher P/N 13-450-953).
Specifications
Material Stainless Steel
Autoclavable Autoclavable
Dimensions (L x W) 24 x 24 in. (61 x 61 cm)
For Use With (Equipment) Metro™ Super Erecta™ shelving system
Length (English) 24 in.
Length (Metric) 61 cm
Width (English) 24 in.
Width (Metric) 61 cm