Learn More
Metro™ Starsys™ Cart Accessories
Product Code. p-4415646
Change view
Click to view available options
Product Type:
Drawer Pull
Unit Size:
Each
Product Code. Product Type unitSize
15927251 Drawer Pull Each
1 options
Product Code. 15927251
Brand: Metro™ SXRDPBK
Parts for use with Starsys carts
  • Black
Specifications
Product Type Drawer Pull
Color Black
For Use With (Equipment) Metro Starsys cart