Learn More
Metro™ Stainless Lab Worktable, Stainless Island Top and Solid HD Shelf
Product Code. 15987091
Click to view available options
Dimensions (L x W x H):
29.75 x 30 x 35 in.
35.75 x 30 x 35 in.
47.75 x 30 x 35 in.
59.75 x 30 x 35 in.
30 x 30 x 35 in.
Includes:
Solid HD Shelf, Adapter Insert
Solid HD shelf
Product Type:
Mobile Table
Stationary Table
Unit Size:
Each
Product Code. 15987091
Brand: Metro™ LTS36IS
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Metro Stainless Lab Worktable features corrosion-proof performance and stainless steel construction throughout.
  • Easily cleaned smooth surfaces
  • Mobile units can be easily moved to clean behind table area
  • Wide range of accessories and caster options available
Order Info
Casters must be ordered separately.
Specifications
Product Type Stationary Table
Dimensions (L x W x H) 35.75 x 30 x 35 in.
For Use With (Application) Ideal for lab
Height (English) 35 in.
Height (Metric) 89 cm
Includes Solid HD shelf
Length (English) 35.75 in.
Length (Metric) 90.8 cm
Load Bearing Capacity (English) 600 lb.
Load Bearing Capacity (Metric) 273 kg
Width (English) 30 in.
Width (Metric) 76.2 cm
Material Stainless Steel