Learn More
Metro™ Stainless Lab Worktable, Gray Phenolic Top and Solid HD Shelf
Product Code. 15917141
Click to view available options
Dimensions (L x W x H):
29.75 x 30 x 36 in.
35.75 x 30 x 36 in.
47.75 x 30 x 35.8 in.
47.75 x 30 x 36 in.
59.75 x 30 x 36 in.
Product Type:
Mobile Lab Worktable
Worktable
Includes:
Solid HD Shelf
Solid HD Shelf, Adapter Insert
Stainless Steel HD Shelf, Adapter Insert, 4 HD Posts
Unit Size:
Each
Product Code. 15917141
Brand: Metro™ LTSM48PG
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Lab worktable provides safety in the laboratory with corrosion-proof performance and stainless steel construction. Features a solid HD shelf and a stainless top/backsplash .
  • Easily cleaned smooth surfaces
  • Mobile units can be easily moved to clean behind table area
  • Wide range of accessories and caster options available
Order Info
Casters must be ordered separately.
Specifications
Product Type Mobile Lab Worktable
Color Silver
Dimensions (L x W x H) 47.75 x 30 x 35.8 in.
For Use With (Application) Laboratory
Height (English) 35.8 in.
Height (Metric) 90.93 cm
Includes Stainless Steel HD Shelf, Adapter Insert, 4 HD Posts
Length (English) 47.75 in.
Length (Metric) 121.28 cm
Load Bearing Capacity (English) 600 lb.
Load Bearing Capacity (Metric) 273 kg
Top Style Gray Phenolic Top or Gray Phenolic Top with Backsplash
Wheel Type Metro 5 in. Stem Casters
Width (English) 30 in.
Width (Metric) 76.2 cm
Material Stainless Steel