Learn More
Metro™ Stainless Lab Worktable, Gray Phenolic Top and 3-Sided Frame
Product Code. 15947101
Change view
Click to view available options
Dimensions (L x W x H):
29.75 x 30 x 36 in.
35.75 x 30 x 36 in.
47.75 x 30 x 36 in.
59.75 x 30 x 36 in.
Product Type:
Mobile Table
Stationary Table
Includes:
3-sided Frame
3-sided Frame, Adaptor Insert
Weight (English):
70 lb.
82 lb.
101 lb.
120 lb.
Unit Size:
Each
Product Code. Dimensions (L x W x H) Product Type Includes Weight (English) unitSize
15947101 35.75 x 30 x 36 in. Stationary Table 3-sided Frame 82 lb. Each
15967091 29.75 x 30 x 36 in. Stationary Table 3-sided Frame 70 lb. Each
15927111 47.75 x 30 x 36 in. Stationary Table 3-sided Frame 101 lb. Each
15907121 59.75 x 30 x 36 in. Stationary Table 3-sided Frame 120 lb. Each
15997121 29.75 x 30 x 36 in. Mobile Table 3-sided Frame, Adaptor Insert 70 lb. Each
15977131 35.75 x 30 x 36 in. Mobile Table 3-sided Frame, Adaptor Insert 82 lb. Each
15957141 47.75 x 30 x 36 in. Mobile Table 3-sided Frame, Adaptor Insert 101 lb. Each
15927151 59.75 x 30 x 36 in. Mobile Table 3-sided Frame, Adaptor Insert 120 lb. Each
8 options
Product Code. 15947101
Brand: Metro™ LTS36UPG
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Lab worktable helps keep workers safe with a phenolic top and corrosion-proof, stainless steel construction throughout the body.
  • Easily cleaned smooth surfaces
  • Mobile units can be easily moved to clean behind table area
  • Wide range of accessories and caster options available
Order Info
Casters must be ordered separately.
Specifications
Product Type Stationary Table
Color Silver
Dimensions (L x W x H) 35.75 x 30 x 36 in.
For Use With (Application) Laboratory
Height (English) 36 in.
Height (Metric) 91.44 cm
Includes 3-sided Frame
Length (English) 35.75 in.
Length (Metric) 90.8 cm
Load Bearing Capacity (English) 600 lb.
Load Bearing Capacity (Metric) 273 kg
Weight (English) 82 lb.
Weight (Metric) 37 kg
Width (English) 30 in.
Width (Metric) 76.2 cm
Material Stainless Steel