Learn More
Metro™ Stainless Lab Worktable, Gray Phenolic Top and 3-Sided Frame
Product Code. 15927111
Click to view available options
Dimensions (L x W x H):
29.75 x 30 x 36 in.
35.75 x 30 x 36 in.
47.75 x 30 x 36 in.
59.75 x 30 x 36 in.
Product Type:
Mobile Table
Stationary Table
Includes:
3-sided Frame
3-sided Frame, Adaptor Insert
Weight (English):
70 lb.
82 lb.
101 lb.
120 lb.
Unit Size:
Each
Product Code. 15927111
Brand: Metro™ LTS48UPG
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Lab worktable helps keep workers safe with a phenolic top and corrosion-proof, stainless steel construction throughout the body.
  • Easily cleaned smooth surfaces
  • Mobile units can be easily moved to clean behind table area
  • Wide range of accessories and caster options available
Order Info
Casters must be ordered separately.
Specifications
Product Type Stationary Table
Color Silver
Dimensions (L x W x H) 47.75 x 30 x 36 in.
For Use With (Application) Laboratory
Height (English) 36 in.
Height (Metric) 91.44 cm
Includes 3-sided Frame
Length (English) 47.75 in.
Length (Metric) 121.2 cm
Load Bearing Capacity (English) 600 lb.
Load Bearing Capacity (Metric) 273 kg
Weight (English) 101 lb.
Weight (Metric) 46 kg
Width (English) 30 in.
Width (Metric) 76.2 cm
Material Stainless Steel