Learn More
Metro™ MetroMax i™ Lab Worktable, Stainless Top and Backsplash with Solid i Shelf
Product Code. 15987061
Click to view available options
Dimensions (L x W x H):
29.75 x 30 x 39.5 in.
Product Type:
Mobile Table
Unit Size:
Each
Product Code. 15987061
Brand: Metro™ LTM30XS3
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The Metro MetroMax i™ Lab Worktable advanced polymer construction provides excellent corrosion-resistance performance for the most corrosive lab environments.
  • Table features smooth surfaces that are easy to clean
  • Mobile units can be easily moved to clean behind table area
  • Available top materials include Type 304 stainless and TRESPA phenolic resin (black or gray)
  • Wide range of accessories and caster options available
Order Info
Casters must be ordered separately.
Specifications
Product Type Mobile Table
Dimensions (L x W x H) 29.75 x 30 x 39.5 in.
For Use With (Application) Ideal for lab
Height (English) 39.5 in.
Height (Metric) 100.3 cm
Includes Backsplash with solid Metro™ MetroMax i™ shelf, adaptor insert
Length (English) 29.75 in.
Length (Metric) 74.9 cm
Load Bearing Capacity (English) 600 lb.
Load Bearing Capacity (Metric) 273 kg
Width (English) 30 in.
Width (Metric) 76.2 cm
Material Polymer