Learn More
Metro™ HD Super Stainless Lab Worktable, Stationary
Product Code. p-175124
Change view
Click to view available options
Dimensions (L x W x H):
48 x 30 x 34 in.
60 x 30 x 34 in.
60 x 36 x 34 in.
70 x 30 x 34 in.
72 x 30 x 34 in.
96 x 30 x 34 in.
Material:
Stainless Steel
Stainless Steel (14 ga.)
Includes:
Bottom 3-sided frame (SS)
Bottom H-frame (SS)
Bottom shelf (SS)
Unit Size:
Each
Product Code. Dimensions (L x W x H) Material Includes unitSize
15967331 60 x 36 x 34 in. Stainless Steel Bottom 3-sided frame (SS) Each
15997321 60 x 30 x 34 in. Stainless Steel Bottom 3-sided frame (SS) Each
15927331 70 x 30 x 34 in. Stainless Steel Bottom 3-sided frame (SS) Each
15957321 48 x 30 x 34 in. Stainless Steel (14 ga.) Bottom shelf (SS) Each
15967321 48 x 30 x 34 in. Stainless Steel (14 ga.) Bottom 3-sided frame (SS) Each
15977321 60 x 30 x 34 in. Stainless Steel (14 ga.) Bottom shelf (SS) Each
15987321 60 x 30 x 34 in. Stainless Steel (14 ga.) Bottom H-frame (SS) Each
15917331 72 x 30 x 34 in. Stainless Steel (14 ga.) Bottom shelf (SS) Each
15947331 96 x 30 x 34 in. Stainless Steel (14 ga.) Bottom shelf (SS) Each
9 options
Product Code. 15967331
Brand: Metro™ WT366US
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Keeping things where the belong in a lab is important to all staff. The Metro HD Super Stainless Lab Worktable is stationary and features an open base that makes it easy to clean. Its stable surface and base construction makes it a choice fit for nearly any lab.
  • 34 in. high (86.4cm) seamless, 14-gauge stainless-steel work surface with rounded edges
  • Support structure virtually eliminates drumming
  • Posts with adjustable feet compensate for slight floor irregularities
  • Leveling foot on each post
Order Info
Shipped knocked down. Assemble easily without tools. Other sizes available; call your Fisher Safety Customer Service Representative at 1-800-772-6733.
Specifications
Product Type Stationary Table
Base Style Open with side and rear crossrails
Certifications/Compliance NSF
Dimensions (L x W x H) 60 x 36 x 34 in.
For Use With (Application) Ideal for lab
Height (English) 34 in.
Height (Metric) 86.4 cm
Includes Bottom 3-sided frame (SS)
Length (English) 60 in.
Length (Metric) 91.4 cm
Weight (English) 104 lb.
Weight (Metric) 47 kg
Width (English) 36 in.
Width (Metric) 91.4 cm
Material Stainless Steel