Learn More
Metro™ HD Super Stainless Lab Worktable, Stationary
Product Code. 15967321
Click to view available options
Dimensions (L x W x H):
48 x 30 x 34 in.
60 x 30 x 34 in.
60 x 36 x 34 in.
70 x 30 x 34 in.
72 x 30 x 34 in.
96 x 30 x 34 in.
Material:
Stainless Steel
Stainless Steel (14 ga.)
Includes:
Bottom 3-sided frame (SS)
Bottom H-frame (SS)
Bottom shelf (SS)
Unit Size:
Each
Product Code. 15967321
Brand: Metro™ WT305US
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Keeping things where the belong in a lab is important to all staff. The Metro HD Super Stainless Lab Worktable is stationary and features an open base that makes it easy to clean. Its stable surface and base construction makes it a choice fit for nearly any lab.
  • 34 in. high (86.4cm) seamless, 14-gauge stainless-steel work surface with rounded edges
  • Support structure virtually eliminates drumming
  • Posts with adjustable feet compensate for slight floor irregularities
  • Leveling foot on each post
Order Info
Shipped knocked down. Assemble easily without tools. Other sizes available; call your Fisher Safety Customer Service Representative at 1-800-772-6733.
Specifications
Product Type Stationary Table
Base Style Open with side and rear crossrails
Certifications/Compliance NSF
Dimensions (L x W x H) 48 x 30 x 34 in.
For Use With (Application) For cleanliness, stability
Frame Material Stainless Steel
Height (English) 34 in.
Height (Metric) 86.36 cm
Includes Bottom 3-sided frame (SS)
Length (English) 48 in.
Length (Metric) 121.92 cm
Weight (English) 130 lb.
Weight (Metric) 58.96 kg
Width (English) 30 in.
Width (Metric) 76.2 cm
Material Stainless Steel (14 ga.)