Learn More
Metro™ Double-Deep Top-Track Stationary Intermediate Unit Kits
missing translation for 'orderingAttributeHoverText'
Width (English):
18 in.
21 in.
24 in.
Width (English) Shelf:
18 in.
21 in.
24 in.
Weight (English):
18 lb.
20 lb.
22 lb.
missing translation for 'unitSize'
Each
Description
- For use with Metro Super Erecta™ and Super Adjustable™ shelving systems
- Chrome-plated finish
- Kit includes connecting hardware
Order Info
One kit is required for intermediate units connected end-to-end. Shelves are sold separately.
Specifications
Specifications
| Product Type | Stationary Intermediate Unit Kit |
| Finish | Chrome |
| For Use With (Equipment) | Metro™ Super Erecta™, Super Adjustable™ shelving system |
| Includes | 86 in. Posts and connecting hardware |
| Weight (English) | 20 lb. |
| Weight (Metric) | 9 kg |
| Width (English) Shelf | 21 in. |
| Width (English) | 21 in. |
| Width (Metric) Shelf | 53 cm |
| Width (Metric) | 53 cm |
| Show More |