Learn More
Metro™ Double-Deep qwikTRAK™ Stationary Intermediate Unit Kits
Connects one double-deep stationary imtermediate unit to to tracks and anchors tracks to floor
10500.00 NOK - 17255.00 NOK
Specifications
Includes | 74 in. High stationary posts and connecting hardware to connect double-deep qwikTRAK™ stationary intermediate units to track and anchors track to floor |
---|---|
Product Type | Stationary Intermediate Unit Kit |
Product Code | Brand | For Use With (Equipment) | Material | Weight (English) | Weight (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Material | Weight (English) | Weight (Metric) | Price | Quantity & Availability | ||
15976971
|
Metro™
LBTAC |
Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ shelving system | Steel | 39.6 lb. | 18 kg |
10500.00 NOK
Each |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15986971
|
Metro™
LBTAK3 |
Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ shelving system | Steel | 39.6 lb. | 18 kg |
12650.00 NOK
Each |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15996971
|
Metro™
LBTAS |
Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ shelving system | Stainless Steel | 39.6 lb. | 18 kg |
17255.00 NOK
Each |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15906981
|
Metro™
LBTAX3 |
Metro™ MetroMax i™ and Q™ shelving system | Polymer | 24.3 lb. | 11 kg |
15410.00 NOK
Each |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
- For use with Metro Super Erecta™, Super Erecta Pro™, Super Adjustable™ and MetroMax iQ shelving systems
- Kit includes 74 in. (188cm) high stationary posts and connecting hardware
Order Info
Shelves are sold separately.
Specifications
74 in. High stationary posts and connecting hardware to connect double-deep qwikTRAK™ stationary intermediate units to track and anchors track to floor | |
Stationary Intermediate Unit Kit |