Learn More
Metro™ Super Erecta™ Heavy Duty Stainless Steel Cantilever Shelf
Product Code. 15906561
Click to view available options
Length (English):
54 in.
60 in.
Material:
Chrome
Stainless Steel
Unit Size:
Each
Product Code. 15906561
Brand: Metro™ 1260CHS
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Heavy duty stainless steel cantilever shelf is a handy and easy way to expand shelving space in Metro™ Super Erecta™ systems.
  • Drop mat design creates a retaining ledge around the entire deep shelf
  • For use with Super Erecta™ or HD Super™ shelves
  • Width: 12 in.
Order Info
Order rear posts that are taller than front posts to accommodate this type of shelf.
Specifications
Product Type Wire Shelf
Dimensions (L x W) 60 x 12 in.
For Use With (Equipment) HD Super™
Length (English) 60 in.
Length (Metric) 152.4 cm
Width (English) 12 in.
Width (Metric) 30.48 cm
Material Stainless Steel