Learn More
Emma Safety Footwear Metric XD Safety Shoes
Product Code. p-7159404
Change view
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. Size unitSize
15938621 37 1 pair
15948621 38 1 pair
15958621 39 1 pair
15968621 40 1 pair
15978621 41 1 pair
15988621 42 1 pair
15998621 43 1 pair
15908631 44 1 pair
15918631 45 1 pair
15928631 46 1 pair
15938631 47 1 pair
15948631 48 1 pair
12 options
Product Code. 15938621
Brand: Emma Safety Footwear MM50756037
Only null left
Add to basket
Add to basket
Unique microfiber upper is fully washable. Features SmartecAir™ for dry, fresh feet. Protective toe cap prevents staining and is lubricant and fat resistant. The sole was developed for use on hard tiled floors and other easy-to-clean floors often seen in high-tech and cleanroom environments.
  • S2-Safety shoe for wet working conditions
  • Microfiber construction
  • GripForce™ Easy Twist Outsole
  • ESD certified
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Design Metric XD
Material Microfiber
Size 37