Learn More
Emma Safety Footwear Metric XD Safety Shoes
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Description
- S2-Safety shoe for wet working conditions
- Microfiber construction
- GripForce™ Easy Twist Outsole
- ESD certified
- Chrome-free lining
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifications
Specifications
| Design | Metric XD |
| Material | Microfiber |
| Size | 47 |