Learn More
SP Bel-Art™ Metric Color Octagon Spinbar Magnetic Stirring Bars
Interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars
Brand:  SP Bel-Art™ F37109-1303
Additional Details : Weight : 0.00570kg
 View more versions of this product
Product Code. 16885383
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • The most commonly used shape.
  • Choose from red, yellow or blue.
  • Pivot ring allows the Spin Bar to “rock” while spinning for better mixing conditions
Specifications
Specifications
3 mm
No Pivot Ring
10 Pk.
Autoclavable
PTFE, FDA-grade, USP Class VI
Blue
PTFE
13 mm