missing translation for 'onlineSavingsMsg'
Learn More

Metoprolol succinate, MedChemExpress

Product Code. 16471866
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16471866

Brand: Medchem Express HY17503A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Metoprolol succinate is an orally active, selective β1-adrenoceptor antagonist. Metoprolol succinate shows anti-inflammation, antitumor and anti-angiogenic properties.

Metoprolol succinate is an orally active, selective β1-adrenoceptor antagonist. Metoprolol succinate shows anti-inflammation, antitumor and anti-angiogenic properties.

Specifications

Chemical Name or Material Metoprolol succinate
CAS 98418-47-4
Color White
Purity Grade Notes Research
Molecular Formula C34H56N2O10
Quantity 500 mg
Solubility Information DMSO : 16.67 mg/mL (25.54 mM; Need ultrasonic)
SMILES OC(CNC(C)C)COC1=CC=C(CCOC)C=C1.OC(CNC(C)C)COC2=CC=C(CCOC)C=C2.OC(CCC(O)=O)=O
Molecular Weight (g/mol) 652.82
Formula Weight 652.82
Percent Purity 99.9%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.