Learn More
Methyltributylphosphoniumiodide, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 1702-42-7 |
| Molecular Formula | C13H30IP |
| MDL Number | MFCD00031626 |
| Quantity | 5 g |
| SMILES | CCCC[P+](CCCC)(C)CCCC.[I-] |
| Molecular Weight (g/mol) | 344.26 |
| Percent Purity | 98% |
| Chemical Name or Material | Methyltributylphosphoniumiodide |