Learn More
Methylphenyl Glyoxylate, Sigma-Aldrich™
Specifications
Specifications
| CAS | 15206-55-0 |
| Melting Point | 17°C (ECHA) |
| Density | 1.16 g/cm3 at 20°C |
| MDL Number | MFCD00008443 |
| Quantity | 25 mL |
| Synonym | Methylphenyl glyoxylate, Phenylglyoxylic acid methyl ester, Benzoyl formic acid methyl ester, Methylphenyl glyoxylate |
| Molecular Weight (g/mol) | 164.16 |
| Grade | Research |