missing translation for 'onlineSavingsMsg'
Learn More

Methyle2-chloro-6-methylpyrimidine-4-carboxylate, 97%, Thermo Scientific™

Product Code. p-2705199
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10427252

Brand: Thermo Scientific Maybridge SPB06334DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 89793-11-3
Molecular Formula C7H7ClN2O2
Molecular Weight (g/mol) 186.595
MDL Number MFCD00117906
InChI Key ZNPORLVPCMFKOR-UHFFFAOYSA-N
Synonym methyl2-chloro-6-methylpyrimidine-4-carboxylate, methyl 2-chloro-4-methylpyrimidine-6-carboxylate, methyl 2-chloro-6-methyl-pyrimidine-4-carboxylate, 4-pyrimidinecarboxylic acid, 2-chloro-6-methyl-, methyl ester, 2-chloro-4-methoxycarbonyl-6-methylpyrimidine, 2-chloro-6-methyl-pyrimidine-4-carboxylic acid methyl ester, acmc-20akio, 2-chloro-4-carbomethoxy-6-methylpyrimidine, methyl 2-chloro-6-methylpyrimidine 4-carboxylate, methyl-2-chloro-6-methylpyrimidine-4-carboxylate
PubChem CID 2736835
IUPAC Name methyl 2-chloro-6-methylpyrimidine-4-carboxylate
SMILES CC1=NC(=NC(=C1)C(=O)OC)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 186.6
Percent Purity 97%
Chemical Name or Material Methyle2-chloro-6-methylpyrimidine-4-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.