Learn More
Macherey-Nagel™ Methylation Smittel TMSH
Product Code. 10123013 Shop All Macherey-Nagel Products
Click to view available options
Quantity:
1 mL
10 mL
Unit Size:
Each
Pack of 20
Product Code. 10123013
Brand: Macherey-Nagel™ 701520.110
Specifications
Item Description Methylation Smittel TMSH
Quantity 10 mL
compliance-icons compliance-icons compliance-icons
Product Identifier
  • TMSH, 0,2 M, 1x10 mL
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : methanol, TMSH