Learn More
Methyl Violet, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| CAS | 8004-87-3 |
| Formula Weight | 393.96 |
| Quantity | 10 g |
| Chemical Name or Material | Methyl Violet |