missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Violet 2B, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 g
Unit Size:
500g
Specifications
Specifications
| CAS | 8004-87-3 |
| Molecular Formula | C24H28ClN3 |
| Synonym | Basic Violet I, Basic Violet K |
| Formula Weight | 393.96 |
| Quantity | 500 g |
| Physical Form | Powder |
| Chemical Name or Material | Methyl Violet 2B |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction