Learn More
(Methyl)triphenylphosphonium Iodide-d3,13CD3, TRC
Specifications
Specifications
| Chemical Name or Material | (Methyl)triphenylphosphonium Iodide-d3,13CD3 |
| CAS | 282107-30-6 |
| Molecular Formula | C1813CH15D3PI |
| Quantity | 25 mg |
| Synonym | (Methyl)triphenyl-phosphonium Iodide-d3,13CD3, Triphenyl(trideuteriomethyl)phosphonium Iodide-13CD3 |
| InChI Formula | InChI=1S/C19H18P.HI/c1-20(17-11-5-2-6-12-17,18-13-7-3-8-14-18)19-15-9-4-10-16-19;/h2-16H,1H3;1H/q+1;/p-1/i1+1D3; |
| SMILES | [2H][13C]([2H])([2H])[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[I-] |
| Molecular Weight (g/mol) | 408.24 |