Learn More
Methyl trans-3-aminocyclohexanecarboxylate hydrochloride, 95%
Product Code. p-7041410
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15446936
Brand: Thermo Scientific Alfa Aesar H62024.MD
Request Bulk or Custom Format
CAS: 712313-64-9 | C8H16ClNO2 | 193.67 g/mol
Methyl trans-3-aminocyclohexanecarboxylate hydrochloride used as catalyst in pharmaceutical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl trans-3-aminocyclohexanecarboxylate hydrochloride used as catalyst in pharmaceutical research.
Solubility
Insoluble in water.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 712313-64-9
Molecular Formula C8H16ClNO2
Molecular Weight (g/mol) 193.67
MDL Number MFCD12022631,MFCD19237735,MFCD22392158,MFCD31620808,MFCD13184930,MFCD28167966
InChI Key OOFXENVWECZJBF-UHFFFAOYNA-N
Synonym trans-methyl-3-aminocyclohexanecarboxylate hydrochloride, trans-methyl-3-aminocyclohexanecarboxylatehydrochloride, methyl trans-3-aminocyclohexanecarboxylate hydrochloride, methyl 1r,3r-3-aminocyclohexane-1-carboxylate hydrochloride
PubChem CID 46172886
SMILES Cl.COC(=O)C1CCCC(N)C1
Specifications
Solubility Information Insoluble in water.
IUPAC Name methyl (1R,3R)-3-aminocyclohexane-1-carboxylate;hydrochloride
Quantity 250 mg
Formula Weight 193.67
Percent Purity 97%
Chemical Name or Material Methyl trans-3-aminocyclohexanecarboxylate hydrochloride