Learn More
Macherey-Nagel™ Methyl-Sil Deactivated Capillary Column
Shop All Macherey-Nagel Products
Click to view available options
Length (Metric):
10 m
25 m
Diameter (Metric) Inner:
0.25 mm
0.32 mm
0.53 mm
Unit Size:
Each
Specifications
Specifications
| Diameter (Metric) | 0.8 mm |
| Type | Deactivated Capillary Column |
| Diameter (Metric) Inner | 0.53 mm |
| For Use With (Application) | Preparation of Capillary Columns Analysis |
| Temperature Range | 320°C |
| Length (Metric) | 10 m |
| Quantity | 1 Each |