Learn More
Methyl Nonadecanoate, SPEX CertiPrep™
Product Code. 11460439 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. 11460439
Brand: SPEX Certiprep S4659
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
AnalyteCAS No.Concentration
Methyl Nonadecanoate1731-94-81000μg/mL
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Packaging 2mL amber prescored glass ampule
CAS 1731-94-8, 67-56-1
CAS Min % 0.1, 99.9
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Physical Form Liquid
Molecular Formula C20H40O2
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI Key BDXAHSJUDUZLDU-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCCCCC(=O)OC
IUPAC Name methyl nonadecanoate
Solvent or Matrix Methanol
Molecular Weight (g/mol) 312.538
PubChem CID 15610
Quantity 1 mL
Chemical Name or Material Methyl Nonadecanoate
Show More Show Less
Product intended for laboratory use only