Learn More
Methyl Nicotinate, Sigma-Aldrich™
Product Code. 15810581
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15810581 500 g 500g
15800581 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15810581 Supplier Sigma Aldrich Supplier No. 8187130500
 Restricted Item
Estimated Shipment: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 93-60-7
Specifications
Melting Point 39°C
Boiling Point 209°C/1013 hPa
MDL Number MFCD00006388
Quantity 500 g
Synonym Methyl nicotinate, Methyl nicotinate
Molecular Weight (g/mol) 137.14
Grade Research