Learn More
Methyl N-(2,3-Dihydro-1,5-dimethyl-3-oxo-2-phenyl-1H-pyrazol-4-yl)-N-methylcarbamate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl N-(2,3-Dihydro-1,5-dimethyl-3-oxo-2-phenyl-1H-pyrazol-4-yl)-N-methylcarbamate |
| CAS | 10116-07-1 |
| Molecular Formula | C14 H17 N3 O3 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C14H17N3O3/c1-10-12(15(2)14(19)20-4)13(18)17(16(10)3)11-8-6-5-7-9-11/h5-9H,1-4H3 |
| SMILES | COC(=O)N(C)C1=C(C)N(C)N(C1=O)c2ccccc2 |
| IUPAC Name | methyl N-(1,5-dimethyl-3-oxo-2-phenylpyrazol-4-yl)-N-methylcarbamate |
| Molecular Weight (g/mol) | 275.303 |
| Formula Weight | 275.127 |