Learn More
Methyl Methacrylate, Sigma-Aldrich™
Product Code. 15848330
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Unit Size:
1 L
100mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15848330 2.5 L 2.5 L
15838330 100 mL 100mL
15806250 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15848330 Supplier Sigma Aldrich Supplier No. 8005902500
 Restricted Item
Estimated Shipment: 18-11-2026
Stabilized for synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 80-62-6
Specifications
Melting Point -48°C
Density 0.94 g/cm3 at 20°C
Boiling Point 101°C/1013 hPa
Molecular Formula CH2=C(CH3)COOCH3
MDL Number MFCD00008587
Quantity 2.5 L
Synonym Methyl methacrylate, Methyl 2-methylpropenoate, Methacrylic acid methyl ester, MMA, Methyl methacrylate
Molecular Weight (g/mol) 100.12
Grade Research