Learn More
Methyl laurate, 98+%, Thermo Scientific Chemicals
Catalog Number p-3758436
Click to view available options
Quantity:
2.5 g
10 g
50 g
250 g
Packaging:
Glass bottle
111-82-0
C13H26O2
214.35
MFCD00008966
UQDUPQYQJKYHQI-UHFFFAOYSA-N
methyl laurate, lauric acid methyl ester, dodecanoic acid, methyl ester, methyl laurinate, methyl dodecylate, methyl n-dodecanoate, uniphat a40, metholene 2296, lauric acid, methyl ester, stepan c40
8139
CHEBI:87494
methyl dodecanoate
CCCCCCCCCCCC(=O)OC
111-82-0
C13H26O2
214.35
MFCD00008966
UQDUPQYQJKYHQI-UHFFFAOYSA-N
methyl laurate, lauric acid methyl ester, dodecanoic acid, methyl ester, methyl laurinate, methyl dodecylate, methyl n-dodecanoate, uniphat a40, metholene 2296, lauric acid, methyl ester, stepan c40
8139
CHEBI:87494
methyl dodecanoate
CCCCCCCCCCCC(=O)OC
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 4°C to 5°C
Color Colorless to Yellow
Density 0.8700g/mL
Boiling Point 261°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)10CO2CH3
Refractive Index 1.4310 to 1.4330
Quantity 2.5 g
Beilstein 02,361
Specific Gravity 0.87
Solubility Information Solubility in water: insoluble
Formula Weight 214.35
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Methyl laurate