Learn More
Methyl isonicotinate, 98%
Product Code. 11420867
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420867 500 g 500g
11400867 25 g 25g
11410867 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11420867 Supplier Thermo Scientific Alfa Aesar Supplier No. B23263.36
Request Bulk or Custom Format
CAS: 2459-09-8 | C7H7NO2 | 137.14 g/mol
Methyl Isonicotinate induces increased walking and take-off behavior in western flower thrips, Frankliniella occidentalis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl Isonicotinate induces increased walking and take-off behavior in western flower thrips, Frankliniella occidentalis.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 9-8-2459
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00006427
InChI Key OLXYLDUSSBULGU-UHFFFAOYSA-N
Synonym methyl isonicotinate, isonicotinic acid methyl ester, 4-pyridinecarboxylic acid, methyl ester, 4-picolinic acid methyl ester, 4-methoxycarbonylpyridine, methylisonicotinate, methyl 4-pyridinecarboxylate, 4-carbomethoxypyridine, isonicotinic acid, methyl ester, i-nicotinic acid, methyl ester
PubChem CID 227085
IUPAC Name methyl pyridine-4-carboxylate
SMILES COC(=O)C1=CC=NC=C1
Specifications
Melting Point 8°C to 8.5°C
Density 1.016
Boiling Point 207°C to 209°C
Flash Point 82°C (180°F)
Odor Characteristic
Refractive Index 1.5135
Quantity 500 g
Solubility Information Slightly soluble in water.
Formula Weight 137.14
Percent Purity 98%
Chemical Name or Material Methyl isonicotinate