Learn More
Methyl Hydrazine-d3 Sulfate, TRC
Product Code. 30467469
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30467469 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30467469 Supplier Toronto Research Chemicals Supplier No. TRCM312302500MG
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl Hydrazine-d3 Sulfate
CAS 70609-01-7
Molecular Formula CH5D3N2O4S
Quantity 500 mg
Synonym MHS-d3, (Methyl-d3)hydrazine Monosulfate, (Methyl-d3)hydrazinium Sulfate, NSC 3802-d3
InChI Formula InChI=1S/CH6N2.H2O4S/c1-3-2;1-5(2,3)4/h3H,2H2,1H3;(H2,1,2,3,4)/i1D3;
SMILES NNC([2H])([2H])[2H].O=S(O)(O)=O
IUPAC Name (methyl-d3)hydrazine sulfate
Molecular Weight (g/mol) 147.17
Formula Weight 147.04