Learn More
Methyl Ethyl Ketone 25L, Solveco
Product Code. 30281142
Change view
Click to view available options
Quantity:
25 L
Unit Size:
25 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30281142 25 L 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30281142 Supplier Solveco Chemicals Supplier No. 1173
  • ONLINE EXCLUSIVE PRICE
    2385.00 NOK
    2170.62 NOK / 25 L
    Save 214.38 NOK
    8% Off
 Restricted Item
Estimated Shipment: 16-11-2026
Methyl ethyl ketone (MEK or Butanone) is a versatile solvent used in processes involving rubber, resins, cellulose acetate, nitrocellulose coatings, and vinyl films.
Methyl ethyl ketone (commonly referred to as MEK or Butanone) is a highly effective solvent capable of dissolving a wide range of substances. It is widely used in industrial processes involving rubber, resins, cellulose acetate, nitrocellulose-based coatings, and vinyl films.
Due to its excellent solvency and fast evaporation rate, MEK is frequently used in the production of plastics, textiles, and paraffin wax. It is also a common component in household and commercial products such as lacquers, varnishes, paint removers, and cleaning agents.
Additionally, MEK is used as a denaturant in the formulation of industrial-grade alcohols.
Specifications
Chemical Name or Material Butan-2-one
CAS 78-93-3
Quantity 25 L
Packaging 15 L HDPE Container
Boiling Point 80 °C
Molecular Weight (g/mol) 72.11 g/mol
Percent Purity 99.90%
Grade Technical