Learn More
Methyl Desoxycholate-d5, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl Desoxycholate-d5 |
| CAS | 52840-15-0 |
| Molecular Formula | C25H37D5O4 |
| Quantity | 5 mg |
| Synonym | (3α,5β,12α)-3,12-Dihydroxy-cholan-2,2,3,4,4-d5-24-oic Acid Methyl Ester, Deoxycholic-d5 Acid Methyl Ester, Methyl 7-Deoxycholate-d5 |
| InChI Formula | InChI=1S/C25H42O4/c1-15(5-10-23(28)29-4)19-8-9-20-18-7-6-16-13-17(26)11-12-24(16,2)21(18)14-22(27)25(19,20)3/h15-22,26-27H,5-14H2,1-4H3/t15-,16-,17-,18?,19-,20+,21?,22+,24+,25-/m1/s1/i11D2,13D2,17D |
| SMILES | O[C@]1([2H])C([2H])([2H])C[C@@]2(C)[C@@](CCC3C2C[C@H](O)[C@@]4(C)[C@@]3([H])CC[C@@H]4[C@@H](CCC(OC)=O)C)([H])C1([2H])[2H] |
| Molecular Weight (g/mol) | 411.63 |
| Formula Weight | 411.3397 |