Learn More
Methyl Cyano(2-nitrophenyl)acetate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl Cyano(2-nitrophenyl)acetate |
| CAS | 113772-13-7 |
| Molecular Formula | C10 H8 N2 O4 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C10H8N2O4/c1-16-10(13)8(6-11)7-4-2-3-5-9(7)12(14)15/h2-5,8H,1H3 |
| SMILES | COC(=O)C(C#N)c1ccccc1[N+](=O)[O-] |
| IUPAC Name | methyl 2-cyano-2-(2-nitrophenyl)acetate |
| Molecular Weight (g/mol) | 220.18 |
| Formula Weight | 220.0484 |