Learn More
Methyl chlorodifluoroacetate, 98%
Product Code. 11488557
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
Product Code. 11488557
Brand: Thermo Scientific Alfa Aesar A18623.09
Request Bulk or Custom Format
CAS: 1514-87-0 | C3H3ClF2O2 | 144.502 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1514-87-0
Molecular Formula C3H3ClF2O2
Molecular Weight (g/mol) 144.502
MDL Number MFCD00000775
InChI Key AWUPLMYXZJKHEG-UHFFFAOYSA-N
Synonym methyl chlorodifluoroacetate, chlorodifluoroacetic acid methyl ester, acetic acid, chlorodifluoro-, methyl ester, methyl difluorochloroacetic acid, methyl chloro difluoro acetate, acetic acid, 2-chloro-2,2-difluoro-, methyl ester, methyl-2-chloro-2,2-difluoroacetate, chloro-difluoro-acetic acid methyl ester, clcf2coome, clcf2co2me
PubChem CID 73935
IUPAC Name methyl 2-chloro-2,2-difluoroacetate
SMILES COC(=O)C(F)(F)Cl
Specifications
Density 1.371
Boiling Point 79°C to 81°C
Flash Point 19°C (66°F)
Refractive Index 1.35
Quantity 10 g
UN Number UN2924
Beilstein 1756072
Sensitivity Moisture sensitive
Formula Weight 144.5
Percent Purity 98%
Chemical Name or Material Methyl chlorodifluoroacetate