missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Benzoate, Supelco™
Description
EMPLURA™ is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.
Specifications
Specifications
| CAS | 93-58-3 |
| Melting Point | -12°C |
| Density | 1.09 g/cm3 at 20°C |
| Boiling Point | 199°C/1013 hPa |
| MDL Number | MFCD00008421 |
| Quantity | 1 L |
| Synonym | Methyl benzoate, Methyl benzoate |
| Molecular Weight (g/mol) | 136.15 |
| Grade | Research |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?