Learn More
Methyl alpha-D-xylopyranoside, 98%, Thermo Scientific Chemicals
Catalog Number 15477749
Click to view available options
Quantity:
1 g
5 g
10 g
91-09-8
C6H12O5
164.157
MFCD00053593
ZBDGHWFPLXXWRD-MOJAZDJTSA-N
methyl alpha-d-xylopyranoside, alpha-d-xylopyranoside, methyl, methyl a-d-xylopyranoside, 2s,3r,4s,5r-2-methoxyoxane-3,4,5-triol, 1-o-methyl-alpha-d-xylopyranoside, beta-methyl-d-xyloside, methyl beta-d-xylopyranoside, methyl-alpha-d-xylopyranoside, alpha methyl-d-xyloside, methyl, a-d-xylopyranoside
101554
(2S,3R,4S,5R)-2-methoxyoxane-3,4,5-triol
COC1C(C(C(CO1)O)O)O
91-09-8
C6H12O5
164.157
MFCD00053593
ZBDGHWFPLXXWRD-MOJAZDJTSA-N
methyl alpha-d-xylopyranoside, alpha-d-xylopyranoside, methyl, methyl a-d-xylopyranoside, 2s,3r,4s,5r-2-methoxyoxane-3,4,5-triol, 1-o-methyl-alpha-d-xylopyranoside, beta-methyl-d-xyloside, methyl beta-d-xylopyranoside, methyl-alpha-d-xylopyranoside, alpha methyl-d-xyloside, methyl, a-d-xylopyranoside
101554
(2S,3R,4S,5R)-2-methoxyoxane-3,4,5-triol
COC1C(C(C(CO1)O)O)O
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Color White
Quantity 1 g
Solubility Information Soluble in water
Optical Rotation +154° (c=10 in Water)
Formula Weight 264.16
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Methyl alpha-D-xylopyranoside