Learn More
Methyl Acetate, Sigma-Aldrich™
Product Code. 15876490
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15876490 1 L 1 L
15869700 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15876490 Supplier Sigma Aldrich Supplier No. 8097111000
 Restricted Item
Estimated Shipment: 19-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 79-20-9
Specifications
Melting Point -98°C
Density 0.93 g/cm3 at 20°C
Boiling Point 56°C to 58°C/1013 hPa
MDL Number MFCD00008711
Quantity 1 L
Molecular Weight (g/mol) 74.08
Grade Research